Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080554.2 | NP_542121.2 | 6, 7 | 862 | ||
GenBank mRNA | AK005335.1 | - | 6, 7 | 840 | ||
AK029067.1 | - | 5, 6 | 1159 | |||
AK032992.1 | - | 6, 7 | 840 | |||
AK078419.1 | - | 6, 7 | 840 | |||
AK145587.1 | - | 6, 7 | 839 | |||
AK148284.1 | - | 863 | ||||
AK205476.1 | - | 1, 2 | 163 | |||
AK207702.1 | - | 1, 2 | 163 | |||
AK220321.1 | - | 6, 7 | 803 | |||
BC019112.1 | - | 6, 7 | 873 |