Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014515.1 | - | 10, 11 | 1238 | ||
AK077621.1 | - | 11, 12 | 1469 | |||
AK088789.1 | - | 11, 12 | 1485 | |||
AK145003.1 | - | 11, 12 | 1474 | |||
BC004596.1 | - | 11, 12 | 1432 | 3584555 | ||
BC004665.1 | - | 1, 2 | 106 |