Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133354.2 | NP_579932.1 | 2 | 227 | ||
GenBank mRNA | AK012619.1 | - | 2 | 228 | ||
AK085238.1 | - | 2 | 174 | |||
AK132213.1 | - | 226 | ||||
AK160404.1 | - | 2 | 244 | |||
AK160418.1 | - | 2 | 246 | |||
AK167308.1 | - | 2 | 237 | |||
BC017522.1 | - | 2 | 201 | 4512364 | ||
BC083326.1 | - | 2 | 139 | 6819700 | ||
BC095930.1 | - | 2 | 214 | 4936947 |