Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 1, 2 | 325 | ||
NM_001010931.2 | NP_001010931.1 | 1, 2 | 325 | |||
NM_001010932.2 | NP_001010932.1 | 1, 2 | 325 | |||
NM_001010933.2 | NP_001010933.1 | 1, 2 | 325 | |||
NM_001010934.2 | NP_001010934.1 | 1, 2 | 325 | |||
XM_006715956.2 | XP_006716019.1 | 2, 3 | 288 | |||
XM_011516115.2 | XP_011514417.1 | 2, 3 | 288 | |||
XM_017012097.1 | XP_016867586.1 | 2, 3 | 288 | |||
XM_017012098.1 | XP_016867587.1 | 2, 3 | 288 | |||
GenBank mRNA | AB208900.1 | - | 1, 2 | 232 | ||
AK313694.1 | - | 1, 2 | 175 | |||
BC022308.1 | - | 1, 2 | 174 | |||
BC063485.1 | - | 1, 2 | 183 | |||
BC105797.1 | - | 1, 2 | 265 | |||
BC130284.1 | - | 1, 2 | 173 | 40146315 | ||
BC130286.1 | - | 1, 2 | 173 | 40146317 | ||
BU533235.1 | - | 264 | ||||
BX495370.1 | - | 1, 2 | 326 | |||
FJ830861.1 | - | 1, 2 | 98 | |||
FJ830862.1 | - | 1, 2 | 98 | |||
L02931.1 | - | 1, 2 | 158 | |||
M29145.1 | - | 1, 2 | 199 | |||
M55379.1 | - | 1, 2 | 98 | |||
M60718.1 | - | 1, 2 | 170 | |||
M73239.1 | - | 1, 2 | 163 | |||
M73240.1 | - | 1, 2 | 163 | |||
M77227.1 | - | 1, 2 | 180 | |||
U46010.1 | - | 1, 2 | 152 | |||
X57574.1 | - | 1, 2 | 175 |