- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Sertad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133210.2 | NP_573473.1 | 2 | 345 | ||
GenBank mRNA | AF317202.1 | - | 2 | 349 | ||
AK134829.1 | - | 2 | 418 | |||
BC034886.1 | - | 2 | 345 | 1364881 |