Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Glcci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286728.1 | NP_001273657.1 | 2 | 98 | ||
NM_001286729.1 | NP_001273658.1 | 2 | 98 | |||
NM_133236.3 | NP_573499.2 | 2 | 774 | |||
GenBank mRNA | AK029524.1 | - | 2 | 100 | ||
AY098636.1 | - | 2 | 820 | |||
BC050802.1 | - | 2 | 136 | 6771009 | ||
CA561411.1 | - | 2 | 154 | |||
CN720421.1 | - | 2 | 131 |