Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pilrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133209.2 | NP_573472.2 | 5 | 827 | ||
GenBank mRNA | AJ400847.1 | - | 5 | 776 | ||
BC145880.1 | - | 5 | 765 | 40131183 | ||
BC145906.1 | - | 5 | 765 | 40131209 |