Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133357.3 | NP_579935.1 | 9 | 2975 | ||
GenBank mRNA | AK028972.1 | - | 9 | 2976 |