Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133667.2 | NP_598428.2 | 4 | 537 | ||
GenBank mRNA | AF267660.1 | - | 4 | 498 | ||
AK002647.1 | - | 4 | 481 | |||
BC021764.1 | - | 4 | 491 | 3488813 | ||
CX223857.1 | - | 4 | 538 |