Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133684.3 | NP_598445.1 | 1 | 215 | ||
GenBank mRNA | AK002874.1 | - | 1 | 213 | ||
AK013440.1 | - | 1 | 235 | |||
AK051230.1 | - | 1 | 216 | |||
AK078650.1 | - | 1 | 216 | |||
AK178160.1 | - | 1 | 41 | |||
AK180823.1 | - | 1 | 41 | |||
BC006888.1 | - | 1 | 198 | 2647757 | ||
BC042686.1 | - | 1 | 219 |