Gene Symbol: Tmem97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133706.2 | NP_598467.1 | 1, 2 | 213 | ||
GenBank mRNA | AK155107.1 | - | 1, 2 | 225 | ||
AK171954.1 | - | 1, 2 | 192 | |||
BC018156.1 | - | 1, 2 | 213 | 4192256 | ||
BC093511.1 | - | 1, 2 | 190 | 5124076 |