Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133743.1 | NP_598504.1 | 2 | 177 | ||
GenBank mRNA | AK009998.1 | - | 2 | 197 | ||
BC003749.1 | - | 2 | 179 | |||
BC016549.1 | - | 2 | 177 | 3585545 | ||
BC094282.1 | - | 2 | 214 | 5029753 |