Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199177.1 | NP_001186106.1 | 2 | 381 | ||
NM_133752.3 | NP_598513.1 | 2 | 381 | |||
GenBank mRNA | AB044138.1 | - | 2 | 373 | ||
AK004715.1 | - | 2 | 336 | |||
AK029157.1 | - | 2 | 339 | |||
AK030835.1 | - | 2 | 335 | |||
AK038446.1 | - | 2 | 328 | |||
AK044657.1 | - | 2 | 314 | |||
AK050383.1 | - | 2 | 321 | |||
AK053861.1 | - | 2 | 339 | |||
AK122311.1 | - | 2 | 309 | |||
AK145620.1 | - | 2 | 333 | |||
BC052704.1 | - | 2 | 323 | |||
BC138665.1 | - | 2 | 262 | 8861686 | ||
BC145959.1 | - | 2 | 262 | 40131262 | ||
CB249413.1 | - | 2 | 382 |