Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133756.4 | NP_598517.1 | 2 | 141 | ||
GenBank mRNA | AK010393.1 | - | 2 | 155 | ||
AK077645.1 | - | 2 | 142 | |||
AK145957.1 | - | 138 | ||||
AK146686.1 | - | 153 | ||||
AK163635.1 | - | 2 | 618 | |||
BC020174.1 | - | 2 | 141 | 4505118 | ||
BC064761.1 | - | 2 | 149 | |||
BC096466.1 | - | 2 | 134 | 3711417 |