Gene Symbol: Smim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133697.3 | NP_598458.1 | 5 | 751 | ||
GenBank mRNA | AK003345.1 | - | 5 | 746 | ||
AK044669.1 | - | 5 | 855 | |||
AK088833.1 | - | 5 | 798 | |||
AK164394.1 | - | 5 | 752 | |||
BC009097.1 | - | 5 | 746 | 3481841 | ||
BC029314.1 | - | 2 | 387 |