Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286007.1 | NP_001272936.1 | 3 | 599 | ||
NM_001286009.1 | NP_001272938.1 | 3 | 599 | |||
NM_001286011.1 | NP_001272940.1 | 3 | 526 | |||
NM_133755.2 | NP_598516.2 | 3 | 526 | |||
GenBank mRNA | AK033157.1 | - | 3 | 316 | ||
AK045084.1 | - | 3 | 315 | |||
AK146771.1 | - | 3 | 521 | |||
AK153511.1 | - | 3 | 448 | |||
AK158060.1 | - | 3 | 316 | |||
AK167924.1 | - | 3 | 421 | |||
BC012519.1 | - | 3 | 454 | 3601589 | ||
BC025564.1 | - | 3 | 448 | |||
BC025582.1 | - | 3 | 415 | 5324981 | ||
BC027034.1 | - | 3 | 510 |