Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833439L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252645.1 | NP_001239574.1 | 5 | 711 | ||
NM_001252646.1 | NP_001239575.1 | 4 | 604 | |||
NM_001252647.1 | NP_001239576.1 | 3 | 425 | |||
NM_001252648.1 | NP_001239577.1 | 2 | 298 | |||
NM_133797.4 | NP_598558.1 | 6 | 799 | |||
GenBank mRNA | AK004845.1 | - | 6 | 758 | ||
AK014792.1 | - | 2 | 232 | |||
AK028697.1 | - | 6 | 732 | |||
AK145025.1 | - | 6 | 765 | |||
AK159945.1 | - | 6 | 762 | |||
AK169399.1 | - | 6 | 765 | |||
AK173126.1 | - | 6 | 712 | |||
AK206163.1 | - | 2 | 210 | |||
BC033445.1 | - | 6 | 778 | 4222839 | ||
BC039775.1 | - | 6 | 737 | 4239147 | ||
BC043040.1 | - | 6 | 755 | 5701249 | ||
BC054417.1 | - | 6 | 744 | 3969623 | ||
BQ924175.1 | - | 5 | 663 | |||
BY244215.1 | - | 3 | 388 | |||
BY265900.1 | - | 2 | 299 | |||
CA461006.1 | - | 4 | 606 |
Gene Symbol: Ptrhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204912.1 | NP_001191841.1 | 2 | 1998 | ||
GenBank mRNA | BC067010.1 | - | 3 | 1915 |