Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSC2 SIRNA Assays ›
Gene Symbol:
TSC2
Gene Name:
tuberous sclerosis 2
Gene Aliases:
LAM, PPP1R160, TSC4
Chromosome Location:
Chr.16: 2047804 - 2088720 on Build GRCh38
UniGene ID:
Hs.90303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
8727
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000548.4
NP_000539.2
5
583
NM_001077183.2
NP_001070651.1
5
583
NM_001114382.2
NP_001107854.1
5
583
NM_001318827.1
NP_001305756.1
4
472
NM_001318829.1
NP_001305758.1
4
416
NM_001318832.1
NP_001305761.1
5
484
XM_005255529.4
XP_005255586.2
5
470
XM_005255531.4
XP_005255588.2
5
470
XM_011522636.2
XP_011520938.1
5
470
XM_011522637.2
XP_011520939.1
5
470
XM_011522638.2
XP_011520940.2
4
563
XM_011522639.2
XP_011520941.1
5
470
XM_011522640.2
XP_011520942.1
5
470
XM_017023615.1
XP_016879104.1
5
470
XM_017023616.1
XP_016879105.1
5
470
XM_017023617.1
XP_016879106.1
4
563
XM_017023618.1
XP_016879107.1
5
470
GenBank mRNA
AB210000.1
-
5
489
AK295672.1
-
4
303
AK295728.1
-
5
484
AK299343.1
-
4
353
BC150300.1
-
5
458
8860381
BF194994.1
-
2
90
DA316794.1
-
5
499
KJ534954.1
-
4
383
KJ534955.1
-
4
383
KJ534958.1
-
4
383
KJ534982.1
-
4
383
KJ534984.1
-
4
383
KJ535011.1
-
4
383
KJ535038.1
-
4
1188
KJ535039.1
-
4
383
KJ535091.1
-
4
383
X75621.1
-
4
401
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
negative regulation of protein kinase activity
protein import into nucleus
endocytosis
cell cycle arrest
heart development
protein localization
negative regulation of cell proliferation
negative regulation of phosphatidylinositol 3-kinase signaling
viral process
vesicle-mediated transport
regulation of endocytosis
negative regulation of Wnt signaling pathway
negative regulation of TOR signaling
protein kinase B signaling
positive regulation of GTPase activity
regulation of insulin receptor signaling pathway
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
positive chemotaxis
regulation of small GTPase mediated signal transduction
regulation of cell cycle
negative regulation of protein kinase B signaling
GTPase activator activity
protein binding
phosphatase binding
small GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0