Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168281.1 | NP_001161753.1 | 7 | 1782 | ||
NM_133784.3 | NP_598545.2 | 7 | 1861 | |||
GenBank mRNA | AJ299430.1 | - | 7 | 1796 | ||
AK009983.1 | - | 7 | 1792 | |||
AK142299.1 | - | 7 | 1783 | |||
AK142720.1 | - | 7 | 1785 |