Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133791.4 | NP_598552.2 | 19 | 3240 | ||
GenBank mRNA | AB045323.1 | - | 19 | 3265 | ||
AK028956.1 | - | 857 | ||||
AK032989.1 | - | 15 | 2306 | |||
AK079375.1 | - | 19 | 3076 | |||
AK147648.1 | - | 3234 | ||||
BC054434.1 | - | 9 | 1521 | |||
BC067050.1 | - | 19 | 3240 | 30537299 |