Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133823.4 | NP_598584.2 | 2 | 319 | ||
GenBank mRNA | AK050255.1 | - | 2 | 301 | ||
AK160235.1 | - | 3 | 649 | |||
BC021954.1 | - | 1 | 270 | 5134853 | ||
BC085229.1 | - | 1 | 282 |