Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134023.1 | NP_598784.1 | 3 | 445 | ||
GenBank mRNA | AK140872.1 | - | 3 | 457 | ||
AK153677.1 | - | 427 | ||||
AK197604.1 | - | 1 | 37 | |||
AK220243.1 | - | 456 | ||||
BC018300.1 | - | 3 | 445 | 4193046 | ||
BC037230.1 | - | 3 | 428 | 2650923 |