Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134054.2 | NP_598815.2 | 2 | 465 | ||
GenBank mRNA | AK003171.1 | - | 2 | 402 | ||
AK003275.1 | - | 2 | 406 | |||
AK008904.1 | - | 2 | 422 | |||
AK018380.1 | - | 2 | 442 | |||
AK137543.1 | - | 2 | 440 | |||
BC022674.1 | - | 2 | 391 | 4236922 |