Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170430.1 | NP_001163901.1 | 2, 3 | 424 | ||
NM_001170431.1 | NP_001163902.1 | 2, 3 | 559 | |||
NM_134060.4 | NP_598821.1 | 2, 3 | 402 | |||
GenBank mRNA | AK076592.1 | - | 2, 3 | 397 | ||
AK077208.1 | - | 2, 3 | 378 | |||
AK141280.1 | - | 2, 3 | 560 | |||
BB851850.1 | - | 2, 3 | 399 | |||
BC006881.1 | - | 2, 3 | 416 | 3596363 |