Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134054.2 | NP_598815.2 | 2 | 518 | ||
GenBank mRNA | AK003171.1 | - | 2 | 455 | ||
AK003275.1 | - | 2 | 459 | |||
AK008904.1 | - | 2 | 475 | |||
AK018380.1 | - | 2 | 495 | |||
AK137543.1 | - | 2 | 493 | |||
BC022674.1 | - | 2 | 444 | 4236922 |