Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133966.2 | NP_598727.1 | 4, 5 | 1104 | ||
GenBank mRNA | AK003477.1 | - | 1, 2 | 230 | ||
AK033477.1 | - | 4, 5 | 1132 | |||
AK089463.1 | - | 4, 5 | 1184 | |||
AK154832.1 | - | 4, 5 | 1184 | |||
BC013550.1 | - | 4, 5 | 1104 | 4222283 |