Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000607.2 | NP_000598.2 | 4 | 487 | ||
GenBank mRNA | AK312035.1 | - | 4 | 412 | ||
BC026238.1 | - | 4 | 412 | 4767956 | ||
BC104818.1 | - | 4 | 401 | 8143821 | ||
BC104820.1 | - | 4 | 401 | 8143823 | ||
BC143313.1 | - | 4 | 401 | 9051818 | ||
BC143314.1 | - | 4 | 401 | 9051819 | ||
BT019790.1 | - | 4 | 376 | |||
EU794584.1 | - | 4 | 376 | |||
M13692.1 | - | 4 | 391 | |||
X02544.1 | - | 4 | 454 |