Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037267.1 | - | 1 | 3792 |
Gene Symbol: Adi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134052.2 | NP_598813.1 | 4 | 1365 | ||
GenBank mRNA | AK145831.1 | - | 4 | 1342 | ||
BC005695.1 | - | 4 | 1361 | 3597652 |
Gene Symbol: Snhg17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133696.1 | - | 1 | 2876 |