Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 5 | 408 | ||
NM_134083.4 | NP_598844.3 | 5 | 403 | |||
GenBank mRNA | AA119207.1 | - | 5 | 408 | ||
AK013540.1 | - | 510 | ||||
AK030893.1 | - | 3 | 845 | |||
AK031242.1 | - | 5 | 369 | |||
AK031248.1 | - | 5 | 374 | |||
AK147784.1 | - | 6 | 409 | |||
AK160881.1 | - | 6 | 595 | |||
BC003224.1 | - | 3 | 201 | 3482729 | ||
JN086338.1 | - | 4 | 324 | |||
JN086339.1 | - | 5 | 562 |