Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134193.2 | NP_598954.2 | 1 | 445 | ||
GenBank mRNA | BC132631.1 | - | 1 | 445 | 40130908 |