Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134084.1 | NP_598845.1 | 3 | 325 | ||
GenBank mRNA | AK028349.1 | - | 3 | 342 | ||
BC004041.1 | - | 3 | 325 | 3496285 |