Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000456.2 | NP_000447.2 | 6 | 1840 | ||
NM_001032386.1 | NP_001027558.1 | 5 | 1714 | |||
NM_001032387.1 | NP_001027559.1 | 4 | 1605 | |||
XM_005269112.1 | XP_005269169.1 | 5 | 1696 | |||
XM_017019905.1 | XP_016875394.1 | 7 | 2386 | |||
XM_017019906.1 | XP_016875395.1 | 5 | 1674 | |||
XM_017019907.1 | XP_016875396.1 | 6 | 2455 | |||
XM_017019908.1 | XP_016875397.1 | 4 | 1615 | |||
GenBank mRNA | AK312763.1 | - | 4 | 1579 | ||
BC065193.1 | - | 4 | 1636 | 5227360 | ||
HM013717.1 | - | 3 | 1523 | |||
L31573.1 | - | 3 | 2246 |