Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tymp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138302.1 | NP_612175.1 | 3 | 278 | ||
GenBank mRNA | AB060274.1 | - | 3 | 278 |