Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138606.2 | NP_613072.1 | 2, 3 | 369 | ||
GenBank mRNA | AK043570.1 | - | 2, 3 | 414 | ||
BC027376.1 | - | 2, 3 | 289 | 4954711 | ||
L41495.1 | - | 2, 3 | 369 |