Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000512.4 | NP_000503.1 | 10, 11 | 1236 | ||
NM_001323543.1 | NP_001310472.1 | 9, 10 | 1116 | |||
NM_001323544.1 | NP_001310473.1 | 11, 12 | 1410 | |||
XM_005256301.3 | XP_005256358.1 | 10, 11 | 1236 | |||
XM_011522982.2 | XP_011521284.1 | 11, 12 | 1282 | |||
XM_017023111.1 | XP_016878600.1 | 10, 11 | 3133 | |||
XM_017023112.1 | XP_016878601.1 | 10, 11 | 3133 | |||
XM_017023113.1 | XP_016878602.1 | 9, 10 | 1094 | |||
GenBank mRNA | AK021488.1 | - | 3, 4 | 325 | ||
AK131096.1 | - | 8, 9 | 4555 | |||
AK294390.1 | - | 8, 9 | 986 | |||
AK308269.1 | - | 11, 12 | 1266 | |||
AK312655.1 | - | 10, 11 | 1213 | |||
AY129021.1 | - | 8, 9 | 913 | |||
BC007825.2 | - | 3, 4 | 296 | |||
BC050684.1 | - | 10, 11 | 1225 | 6091623 | ||
BC056151.1 | - | 10, 11 | 1217 | 6026248 | ||
BE875288.1 | - | 1, 2 | 126 | |||
BP296821.1 | - | 3, 4 | 295 | |||
BX649181.1 | - | 8, 9 | 829 |