Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045863.1 | NP_001039328.1 | 4 | 448 | ||
NM_001045864.1 | NP_001039329.1 | 4 | 534 | |||
NM_138580.2 | NP_613046.2 | 4 | 322 | |||
GenBank mRNA | AK011663.1 | - | 4 | 306 | ||
AK012389.1 | - | 4 | 339 | |||
AK131947.1 | - | 4 | 306 | |||
AK147011.1 | - | 4 | 430 | |||
BC025905.1 | - | 1 | 488 | |||
BC034867.1 | - | 4 | 274 | 1495547 | ||
CN525174.1 | - | 4 | 277 | |||
DV662511.1 | - | 1 | 67 |