Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700088E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138581.2 | NP_613047.1 | 5 | 522 | ||
GenBank mRNA | AK006539.1 | - | 5 | 503 | ||
AK030004.1 | - | 1113 | ||||
BC068203.1 | - | 3 | 384 | |||
BC096672.1 | - | 3 | 437 | 5117646 | ||
BC116891.1 | - | 5 | 453 | 40126210 | ||
BC116893.1 | - | 5 | 453 | 40126212 | ||
BC144714.1 | - | 4 | 270 | 9053262 | ||
BC144715.1 | - | 4 | 382 |