Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138660.2 | NP_619601.2 | 1, 2 | 280 | ||
GenBank mRNA | AF526276.1 | - | 1, 2 | 254 | ||
AJ292072.1 | - | 494 | ||||
AK139608.1 | - | 1, 2 | 319 | |||
BC060672.1 | - | 1, 2 | 108 | |||
BC141296.1 | - | 1, 2 | 393 | 9055908 |