Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139149.2 | NP_631888.1 | 5, 6 | 620 | ||
GenBank mRNA | AF224264.1 | - | 5, 6 | 545 | ||
AK032815.1 | - | 5, 6 | 609 | |||
AK038226.1 | - | 5, 6 | 606 | |||
AK040203.1 | - | 2, 3 | 394 | |||
AK086587.1 | - | 2, 3 | 385 | |||
AK132141.1 | - | 613 | ||||
AK134491.1 | - | 5, 6 | 613 | |||
AK139960.1 | - | 5, 6 | 619 | |||
AK145582.1 | - | 5, 6 | 610 | |||
AK146198.1 | - | 5, 6 | 613 | |||
AK149118.1 | - | 5, 6 | 612 | |||
AK172554.1 | - | 5, 6 | 605 | |||
BC040827.1 | - | 5, 6 | 575 | 4503820 | ||
BC058247.1 | - | 5, 6 | 595 | 6314291 |