Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139200.4 | NP_631939.1 | 1 | 197 | ||
GenBank mRNA | AF192525.1 | - | 1 | 117 | ||
AK030882.1 | - | 1 | 262 | |||
AK037842.1 | - | 1 | 198 | |||
AK089364.1 | - | 1 | 138 | |||
AK152784.1 | - | 1 | 179 | |||
AK157859.1 | - | 1 | 198 | |||
AK164790.1 | - | 2 | 203 | |||
AK166254.1 | - | 1 | 198 | |||
AK168343.1 | - | 1 | 197 | |||
AK169575.1 | - | 1 | 183 | |||
AK170301.1 | - | 1 | 204 | |||
AK170544.1 | - | 1 | 198 | |||
AK170796.1 | - | 1 | 198 | |||
AK171467.1 | - | 1 | 198 | |||
AK172071.1 | - | 1 | 254 | |||
AK172380.1 | - | 1 | 198 | |||
BC007144.1 | - | 1 | 186 | 3487179 |