Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138650.2 | NP_619591.1 | 25 | 2866 | ||
GenBank mRNA | AK220294.1 | - | 25 | 2808 | ||
BC015278.1 | - | 24 | 2439 | 4221290 |