Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 10 | 775 | ||
NM_001166643.1 | NP_001160115.1 | 9 | 689 | |||
NM_138681.4 | NP_619622.3 | 10 | 775 | |||
GenBank mRNA | AF313800.1 | - | 9 | 669 | ||
AK032423.1 | - | 10 | 726 | |||
AK034117.1 | - | 10 | 755 | |||
AK157222.1 | - | 10 | 726 | |||
AK160127.1 | - | 9 | 668 | |||
AK171964.1 | - | 9 | 642 | |||
BC028339.1 | - | 9 | 663 | |||
BC038173.1 | - | 8 | 632 | |||
BC050233.1 | - | 9 | 668 | |||
BC060690.1 | - | 9 | 652 | |||
BC080775.1 | - | 8 | 636 | |||
BC086693.1 | - | 9 | 656 | |||
BC118965.2 | - | 9 | 689 | 40097104 | ||
BC118966.1 | - | 9 | 671 | 40097105 |