Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138756.4 | NP_620095.1 | 7 | 1919 | ||
GenBank mRNA | AK034423.1 | - | 3 | 1455 | ||
AK037383.1 | - | 44 | ||||
AK053646.1 | - | 7 | 2035 | |||
AK078779.1 | - | 7 | 1920 | |||
BC004098.1 | - | 1 | 1112 | |||
BC008171.1 | - | 7 | 1911 | 3710468 |