Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139227.4 | NP_631973.3 | 2, 3 | 561 | ||
GenBank mRNA | AF455111.1 | - | 1, 2 | 301 | ||
AK046679.1 | - | 2, 3 | 562 | |||
BC035459.1 | - | 1, 2 | 214 | |||
BC137846.1 | - | 2, 3 | 482 | 8860865 | ||
CJ101662.1 | - | 2, 3 | 370 |