Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139296.2 | NP_647457.2 | 1 | 161 | ||
GenBank mRNA | AB065134.1 | - | 1 | 161 |