Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lmcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144799.2 | NP_659048.1 | 2, 3 | 312 | ||
GenBank mRNA | AK075847.1 | - | 2, 3 | 313 | ||
AK159946.1 | - | 2, 3 | 263 | |||
BC019124.1 | - | 2, 3 | 263 | 5007525 |