Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144784.3 | NP_659033.1 | 3, 4 | 312 | ||
GenBank mRNA | AJ510150.1 | - | 3, 4 | 304 | ||
AK032097.1 | - | 3, 4 | 283 | |||
AK075593.1 | - | 3, 4 | 282 | |||
AK081715.1 | - | 3, 4 | 312 | |||
AK146241.1 | - | 3, 4 | 267 | |||
AK163402.1 | - | 3, 4 | 316 | |||
AK169771.1 | - | 3, 4 | 305 | |||
BC024429.1 | - | 3, 4 | 315 | |||
BC024763.1 | - | 3, 4 | 284 | 5365469 |