Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139228.3 | NP_631974.1 | 8 | 1149 | ||
GenBank mRNA | AJ313479.1 | - | 8 | 1149 | ||
BC054784.1 | - | 8 | 985 | 6817213 | ||
BC056969.1 | - | 8 | 985 | 6817213 |