Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139272.2 | NP_644678.2 | 1, 2 | 264 | ||
GenBank mRNA | AF348968.1 | - | 1, 2 | 132 | ||
AK046567.1 | - | 1, 2 | 222 | |||
AK145122.1 | - | 1, 2 | 142 | |||
AK149609.1 | - | 1, 2 | 190 | |||
AK151536.1 | - | 1, 2 | 55 | |||
BC059818.1 | - | 1, 2 | 209 | 5697789 | ||
BQ770019.1 | - | 1, 2 | 264 |