Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144802.4 | NP_659051.3 | 11 | 1866 | ||
GenBank mRNA | AK012866.1 | - | 11 | 1582 | ||
AK146879.1 | - | 11 | 1828 | |||
AK159075.1 | - | 1 | 610 | |||
BC004763.1 | - | 8 | 839 | |||
BC006805.1 | - | 2 | 1538 | |||
BC012849.1 | - | 11 | 1812 | 3587716 | ||
BC038912.1 | - | 3 | 358 | |||
BC071184.1 | - | 2 | 1532 |